Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NMB
Entrez Gene ID
102142390
Gene Name
neuromedin B
Gene Aliases
-
Chromosome Location
Chr.7: 65127415 - 65130925 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005560395.2 XP_005560452.2 2 - 3 825 96

View Full Product Details