Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC16A8
Entrez Gene ID
102118045
Gene Name
solute carrier family 16 member 8
Gene Aliases
-
Chromosome Location
Chr.10: 13127144 - 13134089 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005567323.2 XP_005567380.1 3 - 4 3125 103

View Full Product Details