Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCGB1A1
Entrez Gene ID
102145185
Gene Name
secretoglobin family 1A member 1
Gene Aliases
-
Chromosome Location
Chr.14: 11865108 - 11868105 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005577595.2 XP_005577652.1 1 - 2 121 57

View Full Product Details