Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BSX
Entrez Gene ID
102128742
Gene Name
brain specific homeobox
Gene Aliases
-
Chromosome Location
Chr.14: 118962588 - 118968223 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005579972.2 XP_005580029.1 2 - 3 679 79

View Full Product Details