Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APOBR
Entrez Gene ID
102121619
Gene Name
apolipoprotein B receptor
Gene Aliases
-
Chromosome Location
Chr.20: 27630164 - 27634453 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005591570.2 XP_005591627.1 2 - 3 2958 72

View Full Product Details