Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAGE1
Entrez Gene ID
102126384
Gene Name
PAGE family member 1
Gene Aliases
-
Chromosome Location
Chr.X: 47983010 - 47990954 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005593566.2 XP_005593623.1 4 - 5 416 75
XM_015443376.1 XP_015298862.1 4 - 5 341 75

View Full Product Details