Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC54
Entrez Gene ID
102144002
Gene Name
coiled-coil domain containing 54
Gene Aliases
-
Chromosome Location
Chr.2: 164812673 - 164814111 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319380.1 NP_001306309.1 1 - 1 1097 73
GenBank mRNA AB179405.1 - 1 - 1 1097 73

View Full Product Details