Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H12D
Entrez Gene ID
102123251
Gene Name
zinc finger CCCH-type containing 12D
Gene Aliases
-
Chromosome Location
Chr.4: 55400419 - 55438094 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005552090.2 XP_005552147.1 3 - 4 1233 91
XM_005552092.2 XP_005552149.1 3 - 4 1185 91
XM_015448948.1 XP_015304434.1 3 - 4 751 91

View Full Product Details