Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM52
Entrez Gene ID
102136767
Gene Name
tripartite motif containing 52
Gene Aliases
-
Chromosome Location
Chr.6: 181317671 - 181323686 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005558844.2 XP_005558901.2 1 - 2 1239 96

View Full Product Details