Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PENK
Entrez Gene ID
102126322
Gene Name
proenkephalin
Gene Aliases
-
Chromosome Location
Chr.8: 57499534 - 57505343 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005563359.2 XP_005563416.1 3 - 4 344 71

View Full Product Details