Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASGR2
Entrez Gene ID
107127693
Gene Name
asialoglycoprotein receptor 2
Gene Aliases
-
Chromosome Location
Chr.16: 7122934 - 7139173 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015437397.1 XP_015292883.1 2 - 3 333 78

View Full Product Details