Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEBOX
Entrez Gene ID
102131319
Gene Name
SEBOX homeobox
Gene Aliases
-
Chromosome Location
Chr.16: 24585126 - 24587626 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005583220.2 XP_005583277.2 1 - 2 203 74

View Full Product Details