Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102114868
Entrez Gene ID
102114868
Gene Name
submaxillary gland androgen-regulated protein 3B
Gene Aliases
SMR3B
Chromosome Location
Chr.5: 64200446 - 64207362 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005555140.2 XP_005555197.1 2 - 3 263 128

View Full Product Details