Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102125617
Entrez Gene ID
102125617
Gene Name
uncharacterized protein CXorf49-like
Gene Aliases
-
Chromosome Location
Chr.X: 68705533 - 68710186 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005593933.2 XP_005593990.1 2 - 3 2302 100

View Full Product Details