Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCE3A
Entrez Gene ID
102136533
Gene Name
late cornified envelope 3A
Gene Aliases
-
Chromosome Location
Chr.1: 99259816 - 99260085 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005541824.2 XP_005541881.1 1 - 1 121 130

View Full Product Details