Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTH2
Entrez Gene ID
102121229
Gene Name
parathyroid hormone 2
Gene Aliases
-
Chromosome Location
Chr.19: 50233997 - 50234913 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005589898.2 XP_005589955.1 2 - 2 265 83

View Full Product Details