Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLX1
Entrez Gene ID
102129817
Gene Name
distal-less homeobox 1
Gene Aliases
-
Chromosome Location
Chr.12: 61022091 - 61026525 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284600.1 NP_001271529.1 5 - 6 907 76
GenBank mRNA AB173282.1 - 5 - 6 907 76

View Full Product Details