Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCNYL2
Entrez Gene ID
102134652
Gene Name
cyclin Y-like 2
Gene Aliases
-
Chromosome Location
Chr.9: 37708182 - 37747705 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287626.1 NP_001274555.1 5 - 6 799 90
XM_005564999.2 XP_005565056.1 5 - 6 806 90
GenBank mRNA AB168587.1 - 5 - 6 799 90

View Full Product Details