Product Details

Assay Reference Genome
Location

Chr.4:149652262 on build GRCm38
Cytoband
4E2
Species
Mus musculus
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
18707
Gene Symbol
Pik3cd
Gene Name
phosphatidylinositol 3-kinase catalytic delta polypeptide
Gene Aliases
2410099E07Rik, 2610208K16Rik, AW545373, p110delta
Location
Chr.4:149649168-149702566 on build GRCm38
Assay Gene Location
Overlaps Intron 25 - Exon 25
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
Pik3cd NM_001029837.2 NP_001025008.2
NM_001164049.1 NP_001157521.1
NM_001164050.1 NP_001157522.1
NM_001164051.1 NP_001157523.1
NM_001164052.1 NP_001157524.1
NM_008840.3 NP_032866.2
AK040867.1 BAC30725.1
AK149935.1 BAE29178.1
AK153304.1
AK171025.1 BAE42194.1
AK172222.1 BAE42890.1
BC035203.1 AAH35203.2
CT010349.1 CAJ18557.1
U86587.1 AAC25676.1

View Full Product Details