Product Details

Assay Reference Genome
Location

Chr.9:59562072 on build GRCm38
Cytoband
9B
Species
Mus musculus
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
15211
Gene Symbol
Hexa
Gene Name
hexosaminidase A
Gene Aliases
Hex-1
Location
Chr.9:59539545-59565109 on build GRCm38
Assay Gene Location
Overlaps Exon 9 - Intron 9
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
Hexa NM_010421.4 NP_034551.2
AK075895.1 BAC36036.1
AK075911.1 BAC36049.1
AK080777.1 BAC38018.1
AK144168.1 BAE25744.1
AK150444.1 BAE29566.1
AK151960.1 BAE30831.1
AK159091.1 BAE34808.1
AK159814.1 BAE35394.1
AK159890.1 BAE35457.1
AK160112.1 BAE35636.1
AK167013.1 BAE39189.1
AK168186.1 BAE40146.1
AK170903.1
AK213356.1
BC010755.1 AAH10755.1
U07631.1 AAA18775.1
X64331.1 CAA45615.1

View Full Product Details