The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007397.2 | NP_031423.1 | 9 - 10 | 1463 | 74 |
GenBank mRNA | AK137432.1 | - | 9 - 10 | 1215 | 74 |
BC106189.1 | - | 10 - 11 | 1448 | 74 | |
M84120.1 | - | 9 - 10 | 1461 | 74 |