The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284380.1 | NP_001271309.1 | 3 - 4 | 1354 | 97 |
NM_001284381.1 | NP_001271310.1 | 4 - 5 | 1504 | 97 | |
NM_007416.4 | NP_031442.2 | 1 - 2 | 1350 | 97 | |
GenBank mRNA | AK004891.1 | - | 4 - 5 | 1461 | 97 |
AK043877.1 | - | 1 - 2 | 1195 | 97 | |
AK139448.1 | - | 1 - 2 | 1353 | 97 | |
BC018330.1 | - | 3 - 4 | 1327 | 97 | |
BC037002.1 | - | 3 - 4 | 1328 | 97 | |
BC138653.1 | - | 1 - 2 | 1068 | 97 | |
BC138654.1 | - | 1 - 2 | 1068 | 97 | |
Y12738.1 | - | 1 - 2 | 1669 | 97 |