Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Birc3
Entrez Gene ID
11796
Gene Name
baculoviral IAP repeat-containing 3
Gene Aliases
AW107670, Api1, Api2, Birc2, HIAP2, IAP1, IAP2, MIAP1, MIAP2, MIHB, MIHC, RNF49, cIAP-1, cIAP-2, cIAP1, cIAP2
Chromosome Location
Chr.9: 7848699 - 7873213 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007464.3 NP_031490.2 5 - 6 1523 60
GenBank mRNA AK154721.1 - 4 - 5 1188 60
AK156097.1 - 3 - 4 2795 60
AK162108.1 - 5 - 6 1314 60
AK172636.1 - 5 - 6 1256 60
U88908.1 - 5 - 6 1379 60

View Full Product Details