The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007465.2 | NP_031491.2 | 3 - 4 | 1876 | 65 |
GenBank mRNA | BC033480.1 | - | 3 - 4 | 1890 | 65 |
BC145985.1 | - | 3 - 4 | 1729 | 65 | |
L49433.1 | - | 3 - 4 | 1571 | 65 | |
U88909.1 | - | 3 - 4 | 1877 | 65 |