The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007473.4 | NP_031499.1 | 7 - 8 | 1139 | 71 |
GenBank mRNA | AB010100.1 | - | 7 - 8 | 1030 | 71 |
AB056091.1 | - | 7 - 8 | 973 | 71 | |
AK076642.1 | - | 7 - 8 | 1107 | 71 | |
AK131744.1 | - | 7 - 8 | 1107 | 71 | |
BC022223.1 | - | 5 - 6 | 769 | 71 |