The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007479.3 | NP_031505.1 | 3 - 4 | 480 | 112 |
GenBank mRNA | AK002302.1 | - | 3 - 4 | 503 | 112 |
AK013892.1 | - | 3 - 4 | 535 | 112 | |
AK081686.1 | - | 3 - 4 | 480 | 112 | |
AK153011.1 | - | 3 - 4 | 464 | 112 | |
AK168793.1 | - | 3 - 4 | 501 | 112 | |
AK208695.1 | - | 1 - 2 | 99 | 112 | |
BC115650.1 | - | 3 - 4 | 359 | 112 | |
BC115651.1 | - | 3 - 4 | 359 | 112 | |
D87901.1 | - | 3 - 4 | 297 | 112 |