The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007551.2 | NP_031577.2 | 1 - 2 | 104 | 126 |
GenBank mRNA | AK089611.1 | - | 1 - 2 | 117 | 126 |
AK133064.1 | - | 3 - 4 | 336 | 126 | |
AK155954.1 | - | 1 - 2 | 101 | 126 | |
AK156087.1 | - | 1 - 2 | 101 | 126 | |
AK157016.1 | - | 1 - 2 | 101 | 126 | |
BC064059.1 | - | 1 - 2 | 104 | 126 | |
X71788.1 | - | 1 - 2 | 61 | 126 |