Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccnf
Entrez Gene ID
12449
Gene Name
cyclin F
Gene Aliases
CycF, Fbxo1
Chromosome Location
Chr.17: 24223230 - 24251409 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007634.4 NP_031660.3 1 - 2 115 60
GenBank mRNA AK030298.1 - - 54 60
AK031613.1 - 1 - 2 55 60
AK082485.1 - 1 - 2 83 60
AK085183.1 - 1 - 2 52 60
AK159594.1 - 1 - 2 116 60
AK162447.1 - 1 - 2 55 60
AK169263.1 - 1 - 2 66 60
BC037662.1 - 1 - 2 67 60
U20612.1 - 1 - 2 53 60
Z47766.1 - 1 - 2 55 60

View Full Product Details