The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007650.3 | NP_031676.3 | 5 - 6 | 930 | 93 |
GenBank mRNA | AK042105.1 | - | 5 - 6 | 874 | 93 |
AK089076.1 | - | 5 - 6 | 879 | 93 | |
AK143736.1 | - | 5 - 6 | 906 | 93 | |
BC011258.1 | - | 5 - 6 | 930 | 93 | |
M15177.1 | - | 5 - 6 | 808 | 93 | |
U65961.1 | - | 5 - 6 | 808 | 93 |