The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007674.3 | NP_031700.1 | 1 - 2 | 657 | 96 |
GenBank mRNA | AK141681.1 | - | 1 - 2 | 71 | 96 |
BC119158.1 | - | 1 - 2 | 645 | 96 | |
L08061.1 | - | 1 - 2 | 660 | 96 |