The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007711.3 | NP_031737.1 | 8 - 9 | 1938 | 109 |
NM_173873.1 | NP_776298.1 | 9 - 10 | 2114 | 109 | |
NM_173874.1 | NP_776299.1 | 9 - 10 | 2114 | 109 | |
NM_173876.3 | NP_776301.1 | 8 - 9 | 1659 | 109 | |
GenBank mRNA | AF029347.1 | - | 8 - 9 | 1562 | 109 |
AK141136.1 | - | 9 - 10 | 2052 | 109 | |
AK147364.1 | - | 8 - 9 | 2094 | 109 | |
AK147548.1 | - | 8 - 9 | 2096 | 109 | |
AK147621.1 | - | 8 - 9 | 2096 | 109 | |
AK149276.1 | - | 9 - 10 | 1653 | 109 | |
AK150688.1 | - | 9 - 10 | 2043 | 109 | |
AK159698.1 | - | 9 - 10 | 2088 | 109 | |
AK161327.1 | - | 9 - 10 | 2042 | 109 | |
AK162639.1 | - | 9 - 10 | 2055 | 109 | |
AK169297.1 | - | 9 - 10 | 2035 | 109 | |
BC057133.1 | - | 8 - 9 | 1592 | 109 | |
BC057855.1 | - | 8 - 9 | 1754 | 109 | |
X78874.1 | - | 8 - 9 | 1763 | 109 |