Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Csk
Entrez Gene ID
12988
Gene Name
c-src tyrosine kinase
Gene Aliases
AW212630, p50CSK
Chromosome Location
Chr.9: 57626646 - 57653626 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK136876.1 - 3 - 4 848 80
AK137297.1 - 3 - 4 262 80
AK156058.1 - 3 - 4 400 80
AK170815.1 - 3 - 4 416 80
BC018394.1 - 3 - 4 375 80
BC052006.1 - 3 - 4 433 80
BC095962.1 - 5 - 6 756 80
U05247.1 - 1 - 2 127 80

View Full Product Details