The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007795.1 | NP_031821.1 | 1 - 2 | 46 | 67 |
GenBank mRNA | BC024381.1 | - | 1 - 2 | 38 | 67 |
U18366.1 | - | 1 - 2 | 46 | 67 |