Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cybb
Entrez Gene ID
13058
Gene Name
cytochrome b-245, beta polypeptide
Gene Aliases
C88302, CGD91-phox, Cgd, Cyd, Nox2, gp91-1, gp91phox
Chromosome Location
Chr.X: 9435254 - 9487766 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007807.5 NP_031833.3 3 - 4 328 74
GenBank mRNA AK033570.1 - 3 - 4 272 74
AK053920.1 - 3 - 4 272 74
AK152257.1 - 3 - 4 299 74
AK153154.1 - 3 - 4 299 74
AK153259.1 - 3 - 4 299 74
AK154126.1 - 3 - 4 328 74
AK171526.1 - 3 - 4 314 74
BC042838.1 - 3 - 4 295 74
BC071229.1 - 3 - 4 283 74
FJ168469.1 - 3 - 4 317 74
U43384.1 - 3 - 4 254 74

View Full Product Details