The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007877.1 | NP_031903.1 | 6 - 7 | 1147 | 58 |
GenBank mRNA | BC119286.1 | - | 6 - 7 | 1168 | 58 |
BC137819.1 | - | 6 - 7 | 1168 | 58 | |
X67274.1 | - | 6 - 7 | 1147 | 58 |