The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007910.2 | NP_031936.2 | 3 - 4 | 557 | 77 |
GenBank mRNA | AK012195.1 | - | 3 - 4 | 561 | 77 |
AK142604.1 | - | 3 - 4 | 558 | 77 | |
U90663.1 | - | 3 - 4 | 557 | 77 |