Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aimp1
Entrez Gene ID
13722
Gene Name
aminoacyl tRNA synthetase complex-interacting multifunctional protein 1
Gene Aliases
9830137A06Rik, AIMP1/p43, EMAPII, Emap2, Scye1, p43
Chromosome Location
Chr.3: 132660498 - 132683879 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007926.2 NP_031952.2 1 - 2 87 57
GenBank mRNA AK087932.1 - 1 - 2 42 57
AK133859.1 - 1 - 2 40 57
AK160506.1 - 1 - 2 38 57
BC002054.1 - 1 - 2 42 57
BY256548.1 - 1 - 2 92 57
U10118.1 - 1 - 2 38 57

View Full Product Details