Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fgd1
Entrez Gene ID
14163
Gene Name
FYVE, RhoGEF and PH domain containing 1
Gene Aliases
ZFYVE3
Chromosome Location
Chr.X: 151046118 - 151089686 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008001.4 NP_032027.2 14 - 15 2647 101
GenBank mRNA AK148151.1 - 14 - 15 2648 101
BC004838.1 - 6 - 7 537 101
BC011462.1 - 14 - 15 2584 101
U22325.1 - 14 - 15 2246 101

View Full Product Details