The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008008.4 | NP_032034.1 | 3 - 4 | 847 | 113 |
GenBank mRNA | AK033934.1 | - | 2 - 3 | 1127 | 113 |
AK037172.1 | - | 3 - 4 | 849 | 113 | |
AK086639.1 | - | 3 - 4 | 734 | 113 | |
BC052847.1 | - | 3 - 4 | 730 | 113 | |
Z22703.1 | - | 2 - 3 | 384 | 113 |