Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fgfr4
Entrez Gene ID
14186
Gene Name
fibroblast growth factor receptor 4
Gene Aliases
Fgfr-4
Chromosome Location
Chr.13: 55152818 - 55168759 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008011.2 NP_032037.2 17 - 18 2410 64
GenBank mRNA AK084850.1 - 17 - 18 2410 64
AY493377.2 - 15 - 16 2158 64
BC033313.1 - 17 - 18 2597 64
DQ388428.1 - 16 - 17 2296 64
X59927.1 - 17 - 18 2379 64

View Full Product Details