The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008035.2 | NP_032061.1 | 4 - 5 | 604 | 94 |
GenBank mRNA | BC022108.1 | - | 4 - 5 | 590 | 94 |
BQ176722.1 | - | 1 - 2 | 94 | ||
M64817.1 | - | 5 - 6 | 738 | 94 |