Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fshb
Entrez Gene ID
14308
Gene Name
follicle stimulating hormone beta
Gene Aliases
FSH, Fshbeta
Chromosome Location
Chr.2: 107056140 - 107059651 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008045.2 NP_032071.1 1 - 2 36 83
GenBank mRNA AK017593.1 - 1 - 2 37 83
BC061159.1 - 1 - 2 36 83

View Full Product Details