The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008075.2 | NP_032101.3 | 2 - 3 | 276 | 57 |
GenBank mRNA | AF024620.1 | - | 2 - 3 | 276 | 57 |
BI736525.1 | - | 2 - 3 | 310 | 57 |