Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gcdh
Entrez Gene ID
270076
Gene Name
glutaryl-Coenzyme A dehydrogenase
Gene Aliases
9030411L18, AI266902, D17825, GCD
Chromosome Location
Chr.8: 84886387 - 84893921 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044744.1 NP_001038209.2 3 - 4 334 67
NM_008097.2 NP_032123.3 2 - 3 391 67
GenBank mRNA AK165406.1 - 2 - 3 217 67
AU080952.1 - 3 - 4 191 67
BC061158.1 - 2 - 3 221 67
BG807173.1 - 2 - 3 387 67
BY301880.1 - 3 - 4 335 67
U18992.1 - 2 - 3 172 67

View Full Product Details