The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008113.3 | NP_032139.1 | 1 - 2 | 286 | 99 |
GenBank mRNA | AK144321.1 | - | 1 - 2 | 138 | 99 |
BC002032.1 | - | 1 - 2 | 283 | 99 | |
BC158003.1 | - | 1 - 2 | 151 | 99 | |
L42463.1 | - | 1 - 2 | 117 | 99 | |
U73198.1 | - | 1 - 2 | 126 | 99 |