The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008146.3 | NP_032172.3 | 18 - 19 | 3846 | 64 |
GenBank mRNA | AK032610.1 | - | 2 - 3 | 328 | 64 |
BC043452.1 | - | 10 - 11 | 1708 | 64 | |
BC053002.1 | - | 18 - 19 | 3846 | 64 | |
D78270.2 | - | 18 - 19 | 3652 | 64 |