Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hpn
Entrez Gene ID
15451
Gene Name
hepsin
Gene Aliases
Hlb320
Chromosome Location
Chr.7: 31098725 - 31115326 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110252.2 NP_001103722.1 1 - 2 196 92
NM_008281.4 NP_032307.2 1 - 2 196 92
GenBank mRNA AF030065.1 - 1 - 2 155 92
AK002694.1 - 1 - 2 154 92
AY234104.1 - 1 - 2 155 92
BC138809.1 - 1 - 2 40 92
BC145413.1 - 1 - 2 40 92

View Full Product Details