The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008311.2 | NP_032337.2 | 2 - 3 | 677 | 77 |
GenBank mRNA | AF498254.1 | - | 1 - 2 | 350 | 77 |
AF498255.1 | - | 1 - 2 | 350 | 77 | |
AK033713.1 | - | 2 - 3 | 677 | 77 | |
AY131263.1 | - | 1 - 2 | 140 | 77 | |
AY131264.1 | - | 1 - 2 | 140 | 77 | |
BC023690.1 | - | 2 - 3 | 626 | 77 | |
Z15119.1 | - | 1 - 2 | 385 | 77 |