Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Htr2c
Entrez Gene ID
15560
Gene Name
5-hydroxytryptamine (serotonin) receptor 2C
Gene Aliases
5-HT2C, 5-HT2cR, 5-HTR2C, 5HT1c, Htr1c, SR1
Chromosome Location
Chr.X: 146962513 - 147197277 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008312.4 NP_032338.3 3 - 4 726 77
GenBank mRNA AK034512.1 - 3 - 4 728 77
AK043320.1 - 3 - 4 727 77
AK079648.1 - 3 - 4 728 77
AK086348.1 - 3 - 4 725 77
BC098327.1 - 4 - 5 841 77
BC141085.1 - 3 - 4 305 77
M63685.1 - 3 - 4 589 77
X72230.1 - 3 - 4 590 77

View Full Product Details