The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177973.1 | NP_001171444.1 | 13 - 14 | 2214 | 93 |
NM_001177974.1 | NP_001171445.1 | 12 - 13 | 2094 | 93 | |
NM_001177975.1 | NP_001171446.1 | 12 - 13 | 2094 | 93 | |
NM_001177976.1 | NP_001171447.1 | 12 - 13 | 2094 | 93 | |
NM_008363.2 | NP_032389.2 | 12 - 13 | 2094 | 93 | |
GenBank mRNA | AB088370.1 | - | 12 - 13 | 1650 | 93 |
AF103876.1 | - | 12 - 13 | 2094 | 93 | |
AK045958.1 | - | 12 - 13 | 2003 | 93 | |
AK052933.1 | - | 10 - 11 | 1964 | 93 | |
AK143694.1 | - | 12 - 13 | 2005 | 93 | |
AK144879.1 | - | 12 - 13 | 2003 | 93 | |
AY184362.1 | - | 12 - 13 | 2094 | 93 | |
AY184363.1 | - | 12 - 13 | 2094 | 93 | |
AY184364.1 | - | 12 - 13 | 2094 | 93 | |
BC004778.1 | - | 13 - 14 | 2206 | 93 | |
U56773.2 | - | 12 - 13 | 1918 | 93 |